Learn More
ACROBiosystems Human ACE2 / ACEH Protein, Mouse IgG1 Fc Tag (MALS verified)
Catalog No. 502054818
Encompass_Preferred
Recombinant Protein;50UG;Human ACE2, Mouse IgG1 Fc Tag (AC2-H5205) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Ser 740 (Accession # Q9BYF1-1).;This protein carries a mouse IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 109.8 kDa. The protein migrates as 125-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S protein RBD, His Tag (Cat. No. SPD-C52H3) at 1 μg/mL (100 μL/well) can bind Human ACE2, Mouse IgG1 Fc Tag (Cat. No. AC2-H5205) with a linear range of 0.4-6 ng/mL (QC tested).;Immobilized Human ACE2, Mouse IgG1 Fc Tag (Cat. No. AC2-H5205) at 1 μg/mL (100 μL/well) can bind SARS-CoV-2 S1 protein, Fc Tag (Cat. No. S1N-C5255) with a linear range of 1-20 ng/mL (Routinely tested).
Catalog No. 50-205-4818 Supplier ACROBiosystems Supplier No. AC2H520550UG
Recombinant Protein;50UG;Human ACE2, Mouse IgG1 Fc Tag (AC2-H5205) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Ser 740 (Accession # Q9BYF1-1).;This protein carries a mouse IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 109.8 kDa. The protein migrates as 125-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S protein RBD, His Tag (Cat. No. SPD-C52H3) at 1 μg/mL (100 μL/well) can bind Human ACE2, Mouse IgG1 Fc Tag (Cat. No. AC2-H5205) with a linear range of 0.4-6 ng/mL (QC tested).;Immobilized Human ACE2, Mouse IgG1 Fc Tag (Cat. No. AC2-H5205) at 1 μg/mL (100 μL/well) can bind SARS-CoV-2 S1 protein, Fc Tag (Cat. No. S1N-C5255) with a linear range of 1-20 ng/mL (Routinely tested).