Learn More
Sino Biological Recombinant Human ACE2 Protein (hFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10108H02HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ser 740) of human ACE2 precursor (NP_068576.1) was expressed with the fused Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-224-6576