Learn More
Sino Biological Recombinant Anti-ACE2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10108R015100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ACE2 (rh ACE2; Catalog#10108-H31H; NP_068576.1; Met1-Ser740).
Catalog No. 50-240-9070