Learn More
Sino Biological Human ACE2 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10108R00320
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ACE2 (rh ACE2; Catalog#10108-H31H; NP_068576.1; Met1-Ser740).
Catalog No. 50-237-1947