Learn More
Sino Biological Recombinant Human ACE2 Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 10108H27BB20
Encompass_Preferred
A DNA sequence encoding the human ACE2 (NP_068576.1) (Met1-Ser740) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5475