Learn More
Biomatik Corporation Human Acetyl Coenzyme A Carboxylase Alpha (ACACa) ELISA Kit
Supplier:  Biomatik Corporation EKU02071
Encompass_Preferred
Human Acetyl Coenzyme A Carboxylase Alpha (ACACa) ELISA Kit for detecting ACACa in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13085. Gene ID: 31. Target Synonyms: ACAC; ACC1; ACCA; ACAC; ACC2; ACCB; ACACB; ACACA; Acetyl CoA Carboxylase; Biotin carboxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5604