Learn More
Biomatik Corporation Human Acetylcholinesterase Associated Protein (ACHAP) ELISA Kit
Supplier:  Biomatik Corporation EKU02078
Encompass_Preferred
Human Acetylcholinesterase Associated Protein (ACHAP) ELISA Kit for detecting ACHAP in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.051ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60888. Gene ID: 51596. Target Synonyms: CUTA; C6orf82; CutA Divalent Cation Tolerance Homolog; Brain acetylcholinesterase putative membrane anchor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5689