Learn More
Sino Biological Anti-ATP citrate lyase/ACLY Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11769T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ACLY / ATP (rh ACLY / ATP; Catalog#11769-H07B; P53396; Met1-Met1101). ACLY / ATP specific IgG was purified by Human ACLY / ATP affinity chromatography.
Catalog No. 50-255-4560