Learn More
Sino Biological Recombinant Human Aconitase 1/ACO1 Protein (His Tag)
Supplier:  Sino Biological 10966H07B50
Encompass_Preferred
A DNA sequence encoding the human ACO1 (P21399) (Met 1-Lys 889) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-7202