Learn More
Sino Biological Anti-ACOT13/THEM2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 15437T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ACOT13/THEM2 (rh ACOT13/THEM2; Catalog#15437-H07E; NP_060943.1; Met1-Asn140). ACOT13/THEM2 specific IgG was purified by Human ACOT13/THEM2 affinity chromatography.
Catalog No. 50-255-4101