Learn More
Sino Biological Human ACOX1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11266RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ACOX1 (rh ACOX1; Catalog#11266-H07B; AAH08767.1; Met1-Leu660). ACOX1 specific IgG was purified by Human ACOX1 affinity chromatography.
Catalog No. 50-237-3698