Learn More
Sino Biological Human ACP5 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10338T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TRACP / ACP5 (rh TRACP / ACP5; Catalog#10550-H08H; NP_001602.1; Met1-Pro320). TRACP / ACP5 specific IgG was purified by Human TRACP / ACP5 affinity chromatography.
Catalog No. 50-237-4119