Learn More
Sino Biological Human ACPP MAb 100ul (Sino Biological)
Supplier:  Sino Biological 90177C07H50
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Prostatic Acid Phosphatase / ACPP (rh Prostatic Acid Phosphatase / ACPP; Catalog#10959-H08H; NP_001127666.1; Met 1-Lys 382). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-4111