Learn More
Sino Biological Human ACRV1 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785530
Encompass_Preferred
The Human ACRV1 ELISA Pair Set is for the quantitative determination of Human ACRV1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for ACRV1 coated on a 96-well plate. Standards and samples are added to the wells, and any ACRV1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-ACRV1 monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of ACRV1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85530 Supplier Sino Biological Supplier No. SEK117895
The Human ACRV1 ELISA Pair Set is for the quantitative determination of Human ACRV1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for ACRV1 coated on a 96-well plate. Standards and samples are added to the wells, and any ACRV1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-ACRV1 monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of ACRV1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.