Learn More
Sino Biological Anti-ACSL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104906T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ACSL1, and purified by antigen affinity chromatography.
Catalog No. 50-241-0952