Learn More
BIOHIPPO Human ACTa1 Microsample
SDP
Catalog No. 5000315030
Encompass
Human ACTa1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ACTa1 in Tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 6 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ACTa1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ACTa1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ACTa1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15030 Supplier BIOHIPPO Supplier No. BHE15211415
Human ACTa1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ACTa1 in Tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 6 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ACTa1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ACTa1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ACTa1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color