Learn More
Sino Biological Anti-ACTA2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100125T4020
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human Alpha-Smooth Muscle Actin (ACTA2), and purified by antigen affinity chromatography.
Catalog No. 50-241-0654