Learn More
Sino Biological Human ACTB MAb 20ul (Sino Biological)
Supplier:  Sino Biological 100166MM1020
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with the N-terminus of the Human beta Actin / ACTB. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-0972