Learn More
American Research Products Inc Human Actin cytoplasmic 2 AC
Catalog No. 5000328555
Encompass
Human Actin cytoplasmic 2 (ACTG1) ELISA kit from CUSABIO Detection Range 25 pg/mL-1600 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28555 Supplier American Research Products Inc Supplier No. CSBEL001222HU
Human Actin cytoplasmic 2 (ACTG1) ELISA kit from CUSABIO Detection Range 25 pg/mL-1600 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich