Learn More
American Research Products Inc Human Actin related protein 2
Catalog No. 5000327848
Encompass
Human Actin-related protein 2/3 compleX subunit 2 (ARPC2) ELISA kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27848 Supplier American Research Products Inc Supplier No. CSBEL002127HU
Human Actin-related protein 2/3 compleX subunit 2 (ARPC2) ELISA kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich