Learn More
Biomatik Corporation Human Actin Related Protein 2/3 Complex Subunit 2 (ARPC2) ELISA Kit
Supplier:  Biomatik Corporation EKU02094
Encompass_Preferred
Human Actin Related Protein 2/3 Complex Subunit 2 (ARPC2) ELISA Kit for detecting ARPC2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15144. Gene ID: 10109. Target Synonyms: ARC34; PNAS-139; p34-Arc; Arp2/3 complex 34 kDa subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6496