Learn More
Biomatik Corporation Human Actinin Alpha 1 (ACTN1) ELISA Kit
Supplier:  Biomatik Corporation EKU10605
Encompass_Preferred
Human Actinin Alpha 1 (ACTN1) ELISA Kit for detecting ACTN1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12814. Gene ID: 87. Target Synonyms: Alpha-actinin cytoskeletal isoform; F-actin cross-linking protein; Non-muscle alpha-actinin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5853