Learn More
American Research Products Inc Human actiVated coagulation fa
Catalog No. 5000326163
Encompass
Human actiVated coagulation factor XII FXIIa ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26163 Supplier American Research Products Inc Supplier No. CSBE09232H
Human actiVated coagulation factor XII FXIIa ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich