Learn More
Biomatik Corporation Human Activated Leukocyte Cell Adhesion Molecule (ALCAM) CLIA Kit
Supplier:  Biomatik Corporation EKU08769
Encompass_Preferred
Human Activated Leukocyte Cell Adhesion Molecule (ALCAM) CLIA Kit for detecting ALCAM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 12.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q13740. Gene ID: 214. Target Synonyms: CD166; MEMD; Activated Leucocyte Cell Adhesion Molecule. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6246