Learn More
Biomatik Corporation Human Activation Induced Cytidine Deaminase (AICDA) ELISA Kit
Supplier:  Biomatik Corporation EKU11807
Encompass_Preferred
Human Activation Induced Cytidine Deaminase (AICDA) ELISA Kit for detecting AICDA in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9GZX7. Gene ID: 57379. Target Synonyms: ARP2; AID; CDA2; HIGM2; Cytidine aminohydrolase; Single-stranded DNA cytosine deaminase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7468