Learn More
Biomatik Corporation Human Activin A Receptor Type I (ACVR1) ELISA Kit
Supplier:  Biomatik Corporation EKU02113
Encompass_Preferred
Human Activin A Receptor Type I (ACVR1) ELISA Kit for detecting ACVR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q04771. Gene ID: 90. Target Synonyms: ACVR1A; ALK2; ACVRLK2; TSRI; FOP; SKR1; ACTRI; Activin receptor-like kinase 2; Serine/threonine-protein kinase receptor R1; TGF-B superfamily receptor type I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5828