Learn More
Invitrogen™ Human Activin A Recombinant Protein
Description
RP-75636 contains a purified 116 amino acid fragment (aa311-426) of mature human Inhibin beta A chain protein with an N-terminal hexahistidine tag. RP-75636 may be used as a positive control in many immunological assays.
Specifications
Specifications
| Accession Number | P08476 |
| Concentration | 0.153 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 0.02 M tris HCl with 1 mM EDTA, 50% glycerol and no preservative; pH 8 |
| Gene ID (Entrez) | 3624 |
| Name | Human Activin A |
| Quantity | 100 μg |
| Source | E. coli |
| Immunogen | N/A |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.