Learn More
Raybiotech Inc Human Activin AB ELISA Kit
Supplier:  Raybiotech Inc ELHACTIVINAB1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 8.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Activin AB.
Catalog No. 50-194-8943