Learn More
Raybiotech Inc Human Activin R2B/ACVR2B ELISA Kit
Supplier:  Raybiotech Inc ELHACVR2B1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 1.3 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Activin R2B.
Catalog No. 50-194-8945