Learn More
ACROBiosystems Human Activin RIA 100ug
Catalog No. 502535530
Encompass_Preferred
Human Activin RIA Protein Fc Tag (ACA-H5254) is expressed from human 293 cells (HEK293). It contains AA Met 21 - Glu 123 (Accession # Q04771).
Catalog No. 50-253-5530 Supplier ACROBiosystems Supplier No. ACAH5254100UG
Human Activin RIA Protein Fc Tag (ACA-H5254) is expressed from human 293 cells (HEK293). It contains AA Met 21 - Glu 123 (Accession # Q04771).