Learn More
ACROBiosystems Human Activin RIIB 200ug
Supplier:  ACROBiosystems ACBH82E3200UG
Encompass_Preferred
Biotinylated Human Activin RIIB Protein HisAvitag (ACB-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Thr 137 (Accession # Q13705-1 ).
Catalog No. 50-253-5526