Learn More
ACROBiosystems ACROBiosystems Human Activin RIIB / ACVR2B Protein, His Tag
Catalog No. 502106261
Encompass_Preferred
Recombinant Protein;1MG;Human Activin RIIB, His Tag (ACB-H5226) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Thr 137 (Accession # Q13705-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.5 kDa. The protein migrates as 22-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6261 Supplier ACROBiosystems Supplier No. ACBH52261MG
Recombinant Protein;1MG;Human Activin RIIB, His Tag (ACB-H5226) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Thr 137 (Accession # Q13705-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.5 kDa. The protein migrates as 22-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.