Learn More
BIOHIPPO Human ACTL6A Microsample
SDP
Catalog No. 5000315736
Encompass
Human ACTL6A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ACTL6A in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 056 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ACTL6A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ACTL6A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ACTL6A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15736 Supplier BIOHIPPO Supplier No. BHE15211344
Human ACTL6A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ACTL6A in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 056 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ACTL6A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ACTL6A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ACTL6A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color