Learn More
Sino Biological Anti-BAF53a/ACTL6A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 105321T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Actin-like protein 6A/ACTL6A/BAF53a, and purified by antigen affinity chromatography.
Catalog No. 50-241-1364