Learn More
Sino Biological Anti-actinin alpha 1/ACTN1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104759T40100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human actinin alpha 1 / ACTN1, and purified by antigen affinity chromatography.
Catalog No. 50-240-9007