Learn More
Sino Biological Anti-ACVR2B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10229RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ACVR2B (rh ACVR2B ; Catalog#10229-H08H; NP_001097.2; Met1-Thr134). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-5423