Learn More
Sino Biological Recombinant Anti-ALK-1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10066R10420
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ALK-1 / ACVRL1 (rh ALK-1 / ACVRL1; Catalog#10066-H08H; NP_000011.2; Met1-Gln118).
Catalog No. 50-241-0397