Learn More
Sino Biological Human ACY1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11066T16100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ACY1 (rh ACY1; Catalog#10549-H08B; NP_000657.1; Met 1-Ser 408). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-4032