Learn More
Sino Biological Anti-ACY3/aminoacylase 3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102759T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human ACY3/aminoacylase 3, and purified by antigen affinity chromatography.
Catalog No. 50-240-8225