Learn More
Biomatik Corporation Human Acyl Coenzyme A Dehydrogenase, Long Chain (ACADL) ELISA Kit
Supplier:  Biomatik Corporation EKU11745
Encompass_Preferred
Human Acyl Coenzyme A Dehydrogenase, Long Chain (ACADL) ELISA Kit for detecting ACADL in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28330. Gene ID: 33. Target Synonyms: LCAD; ACAD4; Long-chain specific acyl-CoA dehydrogenase, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5256