Learn More
Biomatik Corporation Human Acyl Coenzyme A Synthetase Long Chain Family, Member 4 (ACSL4) ELISA Kit
Supplier:  Biomatik Corporation EKU12214
Encompass_Preferred
Human Acyl Coenzyme A Synthetase Long Chain Family, Member 4 (ACSL4) ELISA Kit for detecting ACSL4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60488. Gene ID: 2182. Target Synonyms: ACS4; FACL4; LACS4; MRX63; MRX68; Fatty-Acid-Coenzyme A Ligase,Long-Chain 4; Mental Retardation,X-Linked 63; Lignoceroyl-CoA Synthase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8003