Learn More
Biomatik Corporation Human Acyl Coenzyme A Synthetase Short Chain Family, Member 2 (ACSS2) ELISA Kit
Supplier:  Biomatik Corporation EKU12213
Encompass_Preferred
Human Acyl Coenzyme A Synthetase Short Chain Family, Member 2 (ACSS2) ELISA Kit for detecting ACSS2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NR19. Gene ID: 55902. Target Synonyms: ACS; ACAS2; ACSA; AceCS; MYH7B; Acetyl-Coenzyme A Synthetase 2 (ADP Forming) ; Acyl-activating enzyme. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8004