Learn More
Biomatik Corporation Human Acylglycerol Kinase (AGK) ELISA Kit
Supplier:  Biomatik Corporation EKU02122
Encompass_Preferred
Human Acylglycerol Kinase (AGK) ELISA Kit for detecting AGK in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.081ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q53H12. Gene ID: 55750. Target Synonyms: MULK; Multiple Substrate Lipid Kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6263