Learn More
Sino Biological Human ACYP1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50053RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ACYP1 / Acylphosphatase-1 (rh ACYP1 / Acylphosphatase-1; Catalog#12578-H09E; P07311; Ala 2-Lys 99). ACYP1 / Acylphosphatase-1 specific IgG was purified by Human ACYP1 / Acylphosphatase-1 affinity chromatography.
Catalog No. 50-237-4489