Learn More
Raybiotech Inc Human ADA ELISA Kit
Supplier:  Raybiotech Inc ELHADA1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 21 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human ADA.
Catalog No. 50-194-8947