Learn More
Sino Biological Anti-Adenosine Deaminase Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100833T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Adenosine Deaminase, and purified by antigen affinity chromatography.
Catalog No. 50-255-5500