Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ADAM-12 ELISA Kit

Catalog No. EH10RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH10RBX5 5 x 96 Tests
EH10RB 96 Tests
EH10RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH10RBX5 Supplier Invitrogen™ Supplier No. EH10RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human ADAM-12 quantitates human ADAM-12 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ADAM-12.

ADAM12 is a member of the ADAM (a disintegrin and metalloprotease-like domain) family. Two forms of ADAM12 have been described: ADAM12S and ADAM12L (short and long forms). The short form is a soluble form lacking the transmembrane and cytoplasmic domains. The short form of ADAM12 was reported to provoke myogenesis, and the lack of cytoplasmic domain suggests different regulation pathways (although both forms can be expressed in the same tissue).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43184
Assay Range 0.05-10 ng/mL
Assay Sensitivity 0.05 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias a disintegrin and metallopeptidase domain 12 (meltrin alpha); a disintegrin and metalloprotease domain 12; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 12 (meltrin alpha); ADAM; ADAM 12; ADAM metallopeptidase domain 12; Adam12; ADAM12-OT1; ADAMs; CAR10; Disintegrin and metalloproteinase domain-containing protein 12; MCMP; MCMPMltna; meltrin alpha; meltrin-alpha; metalloendopeptidases; metalloprotease-disintegrin 12 transmembrane; mKIAA4001; MLTN; Mltna; PRO545; RP11-295J3.5; UNQ346; UNQ346/PRO545a disintegrin and metallopeptidase domain 12 (meltrin alpha); a disintegrin and metalloprotease domain 12; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 12 (meltrin alpha); ADAM; ADAM 12; ADAM metallopeptidase domain 12; Adam12; ADAM12-OT1; ADAMs; CAR10; Disintegrin and metalloproteinase domain-containing protein 12; MCMP; MCMPMltna; meltrin alpha; meltrin-alpha; metalloendopeptidases; metalloprotease-disintegrin 12 transmembrane; mKIAA4001; MLTN; Mltna; PRO545; RP11-295J3.5; UNQ346; UNQ346/PRO545;
Gene ID (Entrez) 8038
Gene Symbol ADAM12
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target ADAM12
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.