Learn More
Abcam Human ADAM10 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB3093151X96TESTS
Encompass_Preferred
Human ADAM10 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human ADAM10 with a sensitivity of 5.38 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2468376