Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ADAM12 S Isoform Recombinant Protein

Catalog No. PIRP75523 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP75523 1 mg
PIRP75522 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP75523 Supplier Invitrogen™ Supplier No. RP75523
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

This product contains purified recombinant human ADAM12 S Isoform protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43184
Concentration 0.27 mg/mL
For Use With (Application) Bioactivity, Western Blot
Formulation 25mM sodium acetate with 50% glycerol and no preservative; pH 4.8
Gene ID (Entrez) 8038
Molecular Weight (g/mol) 62.16 kDa
Name Human ADAM12 S Isoform
pH Range 4.8
Purification Method Purified
Quantity 1 mg
Source E. coli
Immunogen N/A
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias a disintegrin and metalloproteinase; ADAM; ADAM 12; ADAM metallopeptidase domain 12; ADAM12; ADAM12-OT1; ADAMs; CAR10; Disintegrin and metalloproteinase domain-containing protein 12; MCMP; MCMPMltna; meltrin-alpha; metalloendopeptidases; metalloprotease-disintegrin 12 transmembrane; MLTN; MLTNA; UNQ346/PRO545
Common Name ADAM12 S Isoform
Gene Symbol ADAM12
Product Type Protein
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Glu208- Ile738 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.