Learn More
Sino Biological Human ADAM15 MAb 100TS (Sino Biological)
Supplier:  Sino Biological 10517R007P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ADAM15 (rh ADAM15; Catalog#10517-H08H; NP_997074.1; Met 1-Thr 696) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-1265